Gene Transcripts
- Gene Symbol
- NCBP3
- Entrez Gene ID
- 55421
- Gene Name
- nuclear cap binding subunit 3
- Gene Aliases
- C17orf85, ELG, HSA277841
- Chromosome Location
- Chr. 17: 3806748 - 3846251 on Build GRCh38
- UniGene ID
- Hs.120963
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114118.2 |
NP_001107590.1
|
12, 13 |
1665 |
|
XM_005256717.2 |
XP_005256774.1
|
12, 13 |
1665 |
|
XM_006721556.2 |
XP_006721619.1
|
11, 12 |
1554 |
|
XM_011523954.2 |
XP_011522256.1
|
12, 13 |
1582 |
|
XM_011523955.2 |
XP_011522257.1
|
11, 12 |
1436 |
|
XM_017024826.1 |
XP_016880315.1
|
11, 12 |
1552 |
|
XM_017024827.1 |
XP_016880316.1
|
6, 7 |
1139 |
|
XM_017024828.1 |
XP_016880317.1
|
7, 8 |
957 |
|
XM_017024829.1 |
XP_016880318.1
|
7, 8 |
957 |
|
GenBank mRNA |
AJ277841.1 |
- |
7, 8 |
962 |
|
AK125048.1 |
- |
9, 10 |
2354 |
|
AK223470.1 |
- |
5, 6 |
841 |
|
BC010707.1 |
- |
5, 6 |
780 |
|
View Full Product Details