Gene Transcripts

Gene Symbol
ASAP3
Entrez Gene ID
55616
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
Gene Aliases
ACAP4, CENTB6, DDEFL1, UPLC1
Chromosome Location
Chr. 1: 23428563 - 23484631 on Build GRCh38
UniGene ID
Hs.437379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143778.1 NP_001137250.1 9 954
NM_017707.3 NP_060177.2 10 981
XM_011541755.1 XP_011540057.1 10 921
XM_017001685.1 XP_016857174.1 10 915
XM_017001686.1 XP_016857175.1 10 921
XM_017001687.1 XP_016857176.1 10 888
XM_017001688.1 XP_016857177.1 10 877
XM_017001689.1 XP_016857178.1 10 938
GenBank mRNA AB056749.1 - 10 902
AK092291.1 - 10 903
AK295100.1 - 9 876
BC060786.1 - 10 929 5298355

View Full Product Details