Gene Transcripts

Gene Symbol
SAR1A
Entrez Gene ID
56681
Gene Name
secretion associated Ras related GTPase 1A
Gene Aliases
SAR1, SARA1, Sara, masra2
Chromosome Location
Chr. 10: 70150204 - 70170529 on Build GRCh38
UniGene ID
Hs.499960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142648.1 NP_001136120.1 6 482
NM_020150.4 NP_064535.1 5 412
GenBank mRNA AF086166.1 - 1 38
AF087897.1 - 5 379
AF217959.1 - 5 379
AF261717.1 - 5 379
AF274026.1 - 5 378
AF318066.1 - 4 279
AK298591.1 - 4 245
AK312469.1 - 5 367
AL136724.1 - 5 395
AY008268.1 - 5 403
BC003658.2 - 5 377 3546972
BG327663.1 - 6 450
CR533492.1 - 4 279
DB216796.1 - 6 482

View Full Product Details