Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020120.3 | NP_064505.1 | 30 | 3485 | |
NR_027671.2 | 30 | 3638 | |||
XM_006712634.3 | XP_006712697.1 | 30 | 3459 | ||
XM_006712635.3 | XP_006712698.1 | 29 | 3429 | ||
XM_006712636.3 | XP_006712699.1 | 30 | 3448 | ||
XM_017004507.1 | XP_016859996.1 | 31 | 3544 | ||
XM_017004508.1 | XP_016859997.1 | 31 | 3499 | ||
XM_017004509.1 | XP_016859998.1 | 16 | 1790 | ||
XM_017004510.1 | XP_016859999.1 | 12 | 2169 | ||
GenBank mRNA | AF227905.1 | - | 30 | 3345 | |
AK023671.1 | - | 11 | 1181 | ||
AK074251.1 | - | 14 | 1509 | ||
AK074376.1 | - | 4 | 480 | ||
AK293066.1 | - | 30 | 3638 | ||
BC041098.1 | - | 30 | 3635 | 5492334 |