Gene Transcripts

Gene Symbol
NMRAL1
Entrez Gene ID
57407
Gene Name
NmrA-like family domain containing 1
Gene Aliases
HSCARG, SDR48A1
Chromosome Location
Chr. 16: 4461677 - 4476320 on Build GRCh38
UniGene ID
Hs.288969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305141.1 NP_001292070.1 4 689
NM_001305142.1 NP_001292071.1 4 676
NM_020677.4 NP_065728.1 4 790
XM_006720905.3 XP_006720968.1 5 727
XM_006720906.2 XP_006720969.1 5 682
XM_011522567.2 XP_011520869.1 4 680
XM_017023485.1 XP_016878974.1 3 653
XM_017023486.1 XP_016878975.1 3 656
XM_017023487.1 XP_016878976.1 3 656
XM_017023488.1 XP_016878977.1 4 765
XM_017023489.1 XP_016878978.1 5 778
XM_017023490.1 XP_016878979.1 4 603
XM_017023491.1 XP_016878980.1 3 656
XM_017023492.1 XP_016878981.1 4 765
GenBank mRNA AF225419.1 - 4 795
AK098784.1 - 4 650
BC002927.2 - 4 570 3954035
BC007364.2 - 4 632 3630659

View Full Product Details