Gene Transcripts

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
Chromosome Location
Chr. 8: 12945674 - 13029777 on Build GRCh38
UniGene ID
Hs.202521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099677.1 NP_001093147.1 2 1502
NM_020844.2 NP_065895.2 5 1792
XM_005273584.4 XP_005273641.1 5 1803
XM_005273585.4 XP_005273642.1 6 1814
XM_005273586.4 XP_005273643.1 6 1848
XM_005273587.4 XP_005273644.1 5 2036
XM_005273588.4 XP_005273645.1 6 1960
XM_005273590.4 XP_005273647.1 4 1492
XM_005273591.4 XP_005273648.1 5 1976
XM_011544596.2 XP_011542898.1 5 2263
XM_011544597.2 XP_011542899.1 5 1615
XM_011544598.2 XP_011542900.1 5 1884
XM_011544600.2 XP_011542902.1 5 1622
XM_011544601.2 XP_011542903.1 5 1745
XM_017013705.1 XP_016869194.1 5 1653
XM_017013706.1 XP_016869195.1 5 1874
XM_017013707.1 XP_016869196.1 6 2019
XM_017013708.1 XP_016869197.1 5 2118
XM_017013709.1 XP_016869198.1 6 2003
XM_017013710.1 XP_016869199.1 7 1962
XM_017013711.1 XP_016869200.1 6 1797
XM_017013712.1 XP_016869201.1 6 2174
XM_017013713.1 XP_016869202.1 5 1918
XM_017013714.1 XP_016869203.1 4 1720
XM_017013715.1 XP_016869204.1 5 2593
GenBank mRNA AB040889.1 - 2 1566
AK307825.1 - 2 1818
BC016633.2 - 2 1145

View Full Product Details