Gene Transcripts

Gene Symbol
MEAF6
Entrez Gene ID
64769
Gene Name
MYST/Esa1 associated factor 6
Gene Aliases
C1orf149, CENP-28, EAF6, NY-SAR-91
Chromosome Location
Chr. 1: 37489960 - 37514819 on Build GRCh38
UniGene ID
Hs.17118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270875.1 NP_001257804.1 3 332
NM_001270876.1 NP_001257805.1 3 332
NM_022756.5 NP_073593.2 3 332
NR_073090.1 3 332
NR_073091.1 3 332
NR_073092.1 3 332
GenBank mRNA AK021792.1 - 3 267
AK225434.1 - 3 267
AK225598.1 - 3 267
AK308118.1 - 3 266
AK309663.1 - 3 293
AY007166.1 - 3 257
AY211926.1 - 3 256
BC016328.1 - 3 291 4102633
BC056406.1 - 3 288 6471256
BM840891.1 - 3 276
BX386498.2 - 3 324
BX538212.1 - 3 268
BX640719.1 - 3 277
DQ099384.1 - 3 259

View Full Product Details