Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023079.4 | NP_075567.2 | 6 | 1011 | |
GenBank mRNA | AB025426.1 | - | 5 | 552 | |
AK023917.1 | - | 5 | 549 | ||
AK024030.1 | - | 7 | 866 | ||
AK091644.1 | - | 4 | 2142 | ||
AK122770.1 | - | 6 | 909 | ||
AK130303.1 | - | 7 | 869 | ||
AK296865.1 | - | 7 | 902 | ||
AL713782.1 | - | 2 | 167 | ||
BC015169.1 | - | 5 | 538 | ||
BC015890.1 | - | 6 | 879 | 3915698 | |
CR457322.1 | - | 5 | 552 | ||
EF623992.1 | - | 6 | 876 |