Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024537.3 | NP_078813.1 | 5 | 575 | |
XM_006719953.2 | XP_006720016.1 | 5 | 655 | ||
XM_011521116.1 | XP_011519418.1 | 6 | 417 | ||
XM_011521117.2 | XP_011519419.1 | 5 | 511 | ||
XM_011521118.2 | XP_011519420.1 | 5 | 511 | ||
XM_017020741.1 | XP_016876230.1 | 4 | 303 | ||
XM_017020742.1 | XP_016876231.1 | 5 | 511 | ||
GenBank mRNA | AB062436.1 | - | 5 | 486 | |
AK022180.1 | - | 5 | 511 | ||
AK301907.1 | - | 4 | 680 | ||
BC007220.1 | - | 5 | 498 | 2822557 | |
DA966145.1 | - | 5 | 530 |