Gene Transcripts
- Gene Symbol
- TXNDC15
- Entrez Gene ID
- 79770
- Gene Name
- thioredoxin domain containing 15
- Gene Aliases
- C5orf14, UNQ335
- Chromosome Location
- Chr. 5: 134873770 - 134901633 on Build GRCh38
- UniGene ID
- Hs.741304
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024715.3 |
NP_078991.3
|
3, 4 |
1407 |
|
GenBank mRNA |
AK026278.1 |
- |
4, 5 |
810 |
|
AK056512.1 |
- |
3, 4 |
1442 |
|
AK092425.1 |
- |
1, 2 |
1472 |
|
AK225454.1 |
- |
4, 5 |
810 |
|
AK225462.1 |
- |
4, 5 |
810 |
|
AK299578.1 |
- |
3, 4 |
700 |
|
AK307529.1 |
- |
2, 3 |
318 |
|
AY358649.1 |
- |
3, 4 |
835 |
|
BC001615.1 |
- |
3, 4 |
894 |
3544923 |
BC032568.1 |
- |
3, 4 |
808 |
5225890 |
BX538131.1 |
- |
3, 4 |
734 |
|
View Full Product Details