Gene Transcripts

Gene Symbol
LRRC8E
Entrez Gene ID
80131
Gene Name
leucine rich repeat containing 8 family member E
Gene Aliases
Chromosome Location
Chr. 19: 7888505 - 7902023 on Build GRCh38
UniGene ID
Hs.501511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001268284.2 NP_001255213.1 4 1707
NM_001268285.2 NP_001255214.1 2 1421
NM_025061.5 NP_079337.2 3 1564
XM_011528319.2 XP_011526621.1 3 2292
GenBank mRNA AK027073.1 - 1 512
AK091134.1 - 3 1564
AL834474.1 - 1 943
BC022216.1 - 1 852
BC070089.1 - 3 1564 30346745
BC108252.1 - 1 673
BX538180.1 - 3 1554

View Full Product Details