Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323550.1 | NP_001310479.1 | 6 | 1684 | |
NM_001323551.1 | NP_001310480.1 | 6 | 1771 | ||
NM_025079.2 | NP_079355.2 | 6 | 1687 | ||
XM_011542198.2 | XP_011540500.1 | 6 | 1767 | ||
GenBank mRNA | AK026884.1 | - | 6 | 1653 | |
AK093343.1 | - | 5 | 1675 | ||
AY920403.1 | - | 5 | 1537 | ||
BC005001.1 | - | 6 | 1611 | 2958199 | |
CR457364.1 | - | 5 | 1537 | ||
CR995909.1 | - | 1 | 63 |