Gene Transcripts
- Gene Symbol
- AMMECR1L
- Entrez Gene ID
- 83607
- Gene Name
- AMMECR1 like
- Gene Aliases
- Chromosome Location
- Chr. 2: 127861630 - 127886408 on Build GRCh38
- UniGene ID
- Hs.546449
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199140.1 |
NP_001186069.1
|
7, 8 |
1094 |
|
NM_031445.2 |
NP_113633.2
|
7, 8 |
1066 |
|
XM_005263804.4 |
XP_005263861.1
|
8, 9 |
1697 |
|
XM_005263806.4 |
XP_005263863.1
|
7, 8 |
1586 |
|
XM_005263807.4 |
XP_005263864.1
|
6, 7 |
1451 |
|
XM_011511952.2 |
XP_011510254.1
|
7, 8 |
1587 |
|
XM_011511954.2 |
XP_011510256.1
|
8, 9 |
1674 |
|
XM_011511955.2 |
XP_011510257.1
|
6, 7 |
616 |
|
GenBank mRNA |
AK095871.1 |
- |
2, 3 |
151 |
|
AK304145.1 |
- |
7, 8 |
1094 |
|
BC016181.1 |
- |
7, 8 |
1066 |
4644159 |
BC078174.1 |
- |
7, 8 |
1076 |
6012672 |
View Full Product Details