Gene Transcripts

Gene Symbol
OTOP2
Entrez Gene ID
92736
Gene Name
otopetrin 2
Gene Aliases
Chromosome Location
Chr. 17: 74922963 - 74933913 on Build GRCh38
UniGene ID
Hs.352515
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178160.2 NP_835454.1 6 1337
XM_011525479.1 XP_011523781.1 7 1547

View Full Product Details