Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136052.2 | NP_001129524.1 | 20 | 2679 | |
NM_001199752.2 | NP_001186681.1 | 20 | 2776 | ||
NM_001199753.1 | NP_001186682.1 | 20 | 2712 | ||
NM_152359.2 | NP_689572.1 | 19 | 2643 | ||
NR_108072.1 | 21 | 2874 | |||
XM_005258505.2 | XP_005258562.1 | 20 | 2749 | ||
XM_005258506.4 | XP_005258563.1 | 19 | 2665 | ||
XM_006723009.3 | XP_006723072.1 | 17 | 2481 | ||
XM_011526438.2 | XP_011524740.1 | 20 | 2805 | ||
XM_011526439.2 | XP_011524741.1 | 20 | 2869 | ||
XM_011526440.2 | XP_011524742.1 | 19 | 2586 | ||
XM_017026265.1 | XP_016881754.1 | 19 | 2633 | ||
XM_017026266.1 | XP_016881755.1 | 19 | 2554 | ||
XM_017026267.1 | XP_016881756.1 | 19 | 2539 | ||
XM_017026268.1 | XP_016881757.1 | 18 | 2382 | ||
XM_017026269.1 | XP_016881758.1 | 18 | 2492 | ||
XM_017026270.1 | XP_016881759.1 | 18 | 2260 | ||
XM_017026271.1 | XP_016881760.1 | 18 | 2349 | ||
XM_017026272.1 | XP_016881761.1 | 18 | 2441 | ||
XM_017026273.1 | XP_016881762.1 | 18 | 2426 | ||
GenBank mRNA | AF357970.1 | - | 19 | 2601 | |
AK074389.1 | - | 12 | 1622 | ||
AK289713.1 | - | 20 | 2768 | ||
AK290092.1 | - | 20 | 2684 | ||
AL831876.1 | - | 8 | 1007 | ||
BC029104.1 | - | 20 | 2643 | 3872727 |