Gene Transcripts

Gene Symbol
OR10X1
Entrez Gene ID
128367
Gene Name
olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Gene Aliases
OR1-13, OR1-14, OR10X1P
Chromosome Location
Chr. 1: 158578919 - 158579899 on Build GRCh38
UniGene ID
Hs.553589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004477.1 NP_001004477.1 1 848

View Full Product Details