Gene Transcripts

Gene Symbol
FUNDC1
Entrez Gene ID
139341
Gene Name
FUN14 domain containing 1
Gene Aliases
Chromosome Location
Chr. X: 44523639 - 44542975 on Build GRCh38
UniGene ID
Hs.7549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173794.3 NP_776155.1 3 378
GenBank mRNA BC035015.2 - 3 262
BC042813.1 - 3 373 5274638
BM837991.1 - 3 404

View Full Product Details