Gene Transcripts
- Gene Symbol
- ASB8
- Entrez Gene ID
- 140461
- Gene Name
- ankyrin repeat and SOCS box containing 8
- Gene Aliases
- PP14212
- Chromosome Location
- Chr. 12: 48147788 - 48157595 on Build GRCh38
- UniGene ID
- Hs.432699
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319296.1 |
NP_001306225.1
|
3, 4 |
445 |
|
NM_001319297.1 |
NP_001306226.1
|
2, 3 |
305 |
|
NM_001319298.1 |
NP_001306227.1
|
3, 4 |
410 |
|
NM_001319299.1 |
NP_001306228.1
|
3, 4 |
406 |
|
NM_001319300.1 |
NP_001306229.1
|
2, 3 |
309 |
|
NM_001319301.1 |
NP_001306230.1
|
2, 3 |
305 |
|
NM_024095.4 |
NP_077000.1
|
2, 3 |
309 |
|
GenBank mRNA |
AF464877.1 |
- |
2, 3 |
201 |
|
AK024908.1 |
- |
2, 3 |
210 |
|
AK289957.1 |
- |
2, 3 |
276 |
|
AK297013.1 |
- |
2, 3 |
214 |
|
BC001321.1 |
- |
2, 3 |
216 |
3456049 |
BM837218.1 |
- |
2, 3 |
309 |
|
BP195070.1 |
- |
2, 3 |
305 |
|
BP210910.1 |
- |
2, 3 |
274 |
|
BP372866.1 |
- |
3, 4 |
406 |
|
CR457336.1 |
- |
1, 2 |
139 |
|
DA071750.1 |
- |
3, 4 |
367 |
|
DA979010.1 |
- |
3, 4 |
327 |
|
View Full Product Details