Gene Transcripts

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
Chromosome Location
Chr. 12: 95943293 - 95968592 on Build GRCh38
UniGene ID
Hs.424907
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152435.2 NP_689648.2 5 844
GenBank mRNA AB075878.1 - 5 799
AK054617.1 - 5 844
AK122802.1 - 5 811
AK290828.1 - 5 799
AL833144.1 - 4 552
BC029146.1 - 5 780 5183113

View Full Product Details