Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144701.2 | NP_653302.2 | 8 | 1071 | |
XM_005270516.2 | XP_005270573.1 | 5 | 717 | ||
XM_011540790.2 | XP_011539092.1 | 8 | 1539 | ||
XM_011540791.2 | XP_011539093.1 | 8 | 1056 | ||
GenBank mRNA | AF461422.1 | - | 8 | 1071 | |
AM990313.1 | - | 5 | 757 | ||
AM990314.1 | - | 4 | 387 | ||
AM990315.1 | - | 4 | 402 | ||
AM990316.1 | - | 4 | 391 | ||
AM990318.1 | - | 5 | 548 | ||
AM990319.1 | - | 3 | 241 | ||
AM990320.1 | - | 3 | 245 | ||
AM990322.1 | - | 2 | 84 | ||
AM990323.1 | - | 5 | 543 | ||
AM990324.1 | - | 5 | 477 | ||
AM990325.1 | - | 5 | 548 | ||
AM990326.1 | - | 4 | 165 | ||
AM990328.1 | - | 4 | 387 | ||
AM990330.1 | - | 3 | 241 | ||
AM990332.1 | - | 4 | 254 | ||
AM990333.1 | - | 3 | 240 | ||
AM990337.1 | - | 5 | 477 | ||
AY937250.1 | - | 6 | 878 | ||
AY937251.1 | - | 5 | 717 | ||
AY937252.1 | - | 5 | 717 | ||
AY937253.1 | - | 6 | 878 | ||
AY937254.1 | - | 6 | 878 | ||
AY937255.1 | - | 6 | 878 | ||
BC016829.1 | - | 3 | 274 | ||
FM212241.1 | - | 6 | 707 |