Gene Transcripts

Gene Symbol
PLEKHG4B
Entrez Gene ID
153478
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
Gene Aliases
Chromosome Location
Chr. 5: 54065 - 189972 on Build GRCh38
UniGene ID
Hs.535800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052909.3 NP_443141.3 14, 15 3029
XM_006714446.3 XP_006714509.1 16, 17 4167
XM_011513968.2 XP_011512270.1 16, 17 4164
XM_011513969.2 XP_011512271.1 16, 17 4044
XM_011513970.1 XP_011512272.1 16, 17 3756
XM_011513971.2 XP_011512273.1 16, 17 4112
XM_017009112.1 XP_016864601.1 16, 17 4168
GenBank mRNA AB067496.2 - 14, 15 3029
AK125163.1 - 3, 4 858

View Full Product Details