Gene Transcripts

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
Chromosome Location
Chr. 18: 54357914 - 54382035 on Build GRCh38
UniGene ID
Hs.208701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288980.1 NP_001275909.1 5 1468
NM_001288981.1 NP_001275910.1 5 1300
NM_001288982.1 NP_001275911.1 3 671
NM_173529.5 NP_775800.3 4 826
NR_110234.1 2 152
XM_005258201.1 XP_005258258.1 4 2169
XM_005258203.1 XP_005258260.1 4 1629
XM_017025567.1 XP_016881056.1 4 1852
GenBank mRNA AB267906.1 - 4 2164
AB330143.1 - 4 1681
AB330144.1 - 4 1624
AB353076.1 - 5 1467
AB353077.1 - 5 1299
AK126503.1 - 3 906
BC036054.2 - 3 728 5273569
BX640877.1 - 4 799
CN359540.1 - 2 156

View Full Product Details