Gene Transcripts
- Gene Symbol
- RNASEH1
- Entrez Gene ID
- 246243
- Gene Name
- ribonuclease H1
- Gene Aliases
- H1RNA, PEOB2, RNH1
- Chromosome Location
- Chr. 2: 3531813 - 3558371 on Build GRCh38
- UniGene ID
- Hs.568006
- Species
- Homo sapiens
- Gene Symbol
- CCDC144NL-AS1
- Entrez Gene ID
- 440416
- Gene Name
- CCDC144NL antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 17: 20868433 - 20905230 on Build GRCh38
- UniGene ID
- Hs.661076
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104185.1 |
|
3 |
820 |
|
GenBank mRNA |
AK057473.1 |
- |
4 |
1080 |
|
View Full Product Details