Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256126.1 | NP_001243055.1 | 3 | 525 | |
NM_203463.2 | NP_982288.1 | 3 | 525 | ||
GenBank mRNA | AK172775.1 | - | 3 | 525 | |
BC109284.1 | - | 3 | 375 | 40031838 | |
BC109285.1 | - | 3 | 375 | 40031840 | |
BE745491.1 | - | 42 | |||
BX393696.2 | - | 3 | 455 |