Gene Transcripts

Gene Symbol
PGBD3
Entrez Gene ID
267004
Gene Name
piggyBac transposable element derived 3
Gene Aliases
Chromosome Location
Chr. 10: 49515105 - 49524281 on Build GRCh38
UniGene ID
Hs.49063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170753.3 NP_736609.2 2 270
GenBank mRNA AK074682.1 - 2 216
BC028954.1 - 2 86 4479639
BC063690.1 - 2 270 4450811
BG121679.1 - 2 269

Gene Symbol
ERCC6
Entrez Gene ID
2074
Gene Name
ERCC excision repair 6, chromatin remodeling factor
Gene Aliases
ARMD5, CKN2, COFS, COFS1, CSB, POF11, RAD26, UVSS1
Chromosome Location
Chr. 10: 49454480 - 49539538 on Build GRCh38
UniGene ID
Hs.49063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277058.1 NP_001263987.1 6 1594
NM_001277059.1 NP_001263988.1 6 1539
GenBank mRNA AK291018.1 - 6 1511
BC034479.1 - 6 1544 4822982

View Full Product Details