Gene Transcripts

Gene Symbol
OR9G4
Entrez Gene ID
283189
Gene Name
olfactory receptor family 9 subfamily G member 4
Gene Aliases
OR11-216
Chromosome Location
Chr. 11: 56742828 - 56743811 on Build GRCh38
UniGene ID
Hs.554521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005284.1 NP_001005284.1 1 664

View Full Product Details