Gene Transcripts

Gene Symbol
LINC00523
Entrez Gene ID
283601
Gene Name
long intergenic non-protein coding RNA 523
Gene Aliases
C14orf70
Chromosome Location
Chr. 14: 100657268 - 100672744 on Build GRCh38
UniGene ID
Hs.379802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024096.1 2 777
GenBank mRNA BC132989.1 - 2 528
BC132991.1 - 2 528
BX161445.1 - 2 777

View Full Product Details