Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289973.1 | NP_001276902.1 | 8 | 1513 | |
NM_207411.4 | NP_997294.3 | 7 | 1375 | ||
XM_011534750.2 | XP_011533052.1 | 3 | 1881 | ||
XM_017013401.1 | XP_016868890.1 | 6 | 1794 | ||
XM_017013402.1 | XP_016868891.1 | 5 | 1586 | ||
GenBank mRNA | AY358489.1 | - | 7 | 1246 | |
AY534242.1 | - | 7 | 1375 | ||
AY534243.1 | - | 8 | 1367 |