Gene Transcripts

Gene Symbol
OR6M1
Entrez Gene ID
390261
Gene Name
olfactory receptor family 6 subfamily M member 1
Gene Aliases
OR11-271
Chromosome Location
Chr. 11: 123805408 - 123806349 on Build GRCh38
UniGene ID
Hs.557924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005325.1 NP_001005325.1 1 24
GenBank mRNA BC136929.1 - 1 62
BC136931.1 - 1 62 9020923

View Full Product Details