Gene Transcripts
- Gene Symbol
- UGT2B17
- Entrez Gene ID
- 7367
- Gene Name
- UDP glucuronosyltransferase family 2 member B17
- Gene Aliases
- BMND12, UDPGT2B17
- Chromosome Location
- Chr. 4: 68537184 - 68568527 on Build GRCh38
- UniGene ID
- Hs.575083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077.3 |
NP_001068.1
|
1 |
417 |
|
GenBank mRNA |
U59209.1 |
- |
1 |
425 |
|
View Full Product Details