Gene Transcripts

Gene Symbol
S1PR4
Entrez Gene ID
8698
Gene Name
sphingosine-1-phosphate receptor 4
Gene Aliases
EDG6, LPC1, S1P4, SLP4
Chromosome Location
Chr. 19: 3178738 - 3180332 on Build GRCh38
UniGene ID
Hs.662006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003775.3 NP_003766.1 1 130
GenBank mRNA AJ000479.1 - 1 97
BC014970.1 - 1 100 4849349

View Full Product Details