Gene Transcripts

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
Chromosome Location
Chr. 1: 174999435 - 175012027 on Build GRCh38
UniGene ID
Hs.508524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007214.1 NP_001007215.1 6 1748
NM_014412.2 NP_055227.1 6 1933
GenBank mRNA AK093425.1 - 6 1720
AL035305.1 - 6 1625

View Full Product Details