Gene Transcripts

Gene Symbol
MAPKAPK5
Entrez Gene ID
8550
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Gene Aliases
MAPKAP-K5, MK-5, MK5, PRAK
Chromosome Location
Chr. 12: 111842228 - 111902225 on Build GRCh38
UniGene ID
Hs.333120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003668.3 NP_003659.2 1 129
NM_139078.2 NP_620777.1 1 129
XM_017020127.1 XP_016875616.1 1 52
XM_017020128.1 XP_016875617.1 1 52
XM_017020131.1 XP_016875620.1 1 52
GenBank mRNA AK122767.1 - 1 379
BC041049.1 - 1 116 4909021
BG502445.1 - 1 130

Gene Symbol
MAPKAPK5-AS1
Entrez Gene ID
51275
Gene Name
MAPKAPK5 antisense RNA 1
Gene Aliases
C12orf47
Chromosome Location
Chr. 12: 111839769 - 111842902 on Build GRCh38
UniGene ID
Hs.333120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015404.1 1 543
GenBank mRNA AK096935.1 - 1 543
BC023547.2 - 1 194
BC136540.1 - 1 468
JQ794500.1 - 1 646

View Full Product Details