Gene Transcripts

Gene Symbol
FXYD7
Entrez Gene ID
53822
Gene Name
FXYD domain containing ion transport regulator 7
Gene Aliases
Chromosome Location
Chr. 19: 35143250 - 35154302 on Build GRCh38
UniGene ID
Hs.134729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022006.1 NP_071289.1 3 204
GenBank mRNA AI929519.1 - 3 204
AK057825.1 - 3 179
BC018619.1 - 3 188 3626060

View Full Product Details