Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001031677.3 | NP_001026847.1 | 1 | 480 | |
NM_130781.3 | NP_570137.2 | 2 | 394 | ||
NR_109789.1 | 2 | 394 | |||
NR_109790.1 | 1 | 480 | |||
GenBank mRNA | AF087904.1 | - | 1 | 198 | |
AF258583.1 | - | 1 | 224 | ||
AK021761.1 | - | 1 | 417 | ||
AK308155.1 | - | 2 | 349 | ||
AL833898.1 | - | 1 | 22 | ||
BC010006.2 | - | 2 | 317 | 4300377 | |
BC015534.1 | - | 2 | 356 | 3916832 | |
BC021263.2 | - | 1 | 440 | 4329216 | |
BT007268.1 | - | 1 | 24 | ||
DA058886.1 | - | 2 | 393 | ||
DA898352.1 | - | 2 | 391 |