Gene Transcripts

Gene Symbol
SLAIN2
Entrez Gene ID
57606
Gene Name
SLAIN motif family member 2
Gene Aliases
KIAA1458
Chromosome Location
Chr. 4: 48341480 - 48426198 on Build GRCh38
UniGene ID
Hs.479677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020846.1 NP_065897.1 5 1051
XM_005248121.2 XP_005248178.1 5 1167
GenBank mRNA AB040891.1 - 5 1021
AK025264.1 - 1 102
AK026476.1 - 5 598
AK291006.1 - 5 1181
BC031691.2 - 3 371
BC040993.1 - 5 1330 5272534

View Full Product Details