Gene Transcripts

Gene Symbol
ARID3C
Entrez Gene ID
138715
Gene Name
AT-rich interaction domain 3C
Gene Aliases
Chromosome Location
Chr. 9: 34621360 - 34633112 on Build GRCh38
UniGene ID
Hs.534549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017363.1 NP_001017363.1 1 297
XM_017014283.1 XP_016869772.1 3 3383

View Full Product Details