Gene Transcripts

Gene Symbol
ATP8B5P
Entrez Gene ID
158381
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
Gene Aliases
FetA
Chromosome Location
Chr. 9: 35406755 - 35483029 on Build GRCh38
UniGene ID
Hs.575661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003582.1 11 1478
NR_110320.1 11 1474
GenBank mRNA BC035229.1 - 10 2884
BC130448.1 - 6 1238
BC130450.1 - 6 1085

View Full Product Details