Gene Transcripts

Gene Symbol
UPF3B
Entrez Gene ID
65109
Gene Name
UPF3 regulator of nonsense transcripts homolog B (yeast)
Gene Aliases
HUPF3B, MRX62, MRXS14, RENT3B, UPF3BP1, UPF3BP2, UPF3BP3, UPF3X, Upf3p-X
Chromosome Location
Chr. X: 119805311 - 119853028 on Build GRCh38
UniGene ID
Hs.103832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017029738.1 XP_016885227.1 15 1969
GenBank mRNA BF223724.1 - 2 188

View Full Product Details