Gene Transcripts
- Gene Symbol
- FAM19A3
- Entrez Gene ID
- 284467
- Gene Name
- family with sequence similarity 19 member A3, C-C motif chemokine like
- Gene Aliases
- TAFA-3, TAFA3
- Chromosome Location
- Chr. 1: 112719003 - 112727235 on Build GRCh38
- UniGene ID
- Hs.439116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001004440.1 |
NP_001004440.1
|
4, 5 |
530 |
|
NM_182759.2 |
NP_877436.1
|
4, 5 |
462 |
|
XM_005270766.3 |
XP_005270823.1
|
5, 6 |
749 |
|
XM_006710582.3 |
XP_006710645.1
|
5, 6 |
817 |
|
GenBank mRNA |
AY325116.1 |
- |
4, 5 |
462 |
|
AY325119.1 |
- |
4, 5 |
530 |
|
BC112268.1 |
- |
4, 5 |
524 |
8327736 |
BC117479.1 |
- |
4, 5 |
524 |
40126030 |
BC143949.1 |
- |
4, 5 |
493 |
|
BC144042.1 |
- |
4, 5 |
513 |
|
BC144043.1 |
- |
4, 5 |
456 |
9052563 |
View Full Product Details