Gene Transcripts
- Gene Symbol
- FRG1
- Entrez Gene ID
- 2483
- Gene Name
- FSHD region gene 1
- Gene Aliases
- FRG1A, FSG1
- Chromosome Location
- Chr. 4: 189940819 - 189963204 on Build GRCh38
- UniGene ID
- Hs.203772
- Species
- Homo sapiens
- Gene Symbol
- FRG1BP
- Entrez Gene ID
- 284802
- Gene Name
- FSHD region gene 1 family member B, pseudogene
- Gene Aliases
- C20orf80, FRG1B, bA348I14.2
- Chromosome Location
- Chr. 20: 30377203 - 30399331 on Build GRCh38
- UniGene ID
- Hs.653099
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003579.1 |
|
5, 6 |
623 |
|
GenBank mRNA |
BC062743.1 |
- |
6 |
842 |
|
BC095491.1 |
- |
5, 6 |
601 |
|
- Gene Symbol
- FRG1CP
- Entrez Gene ID
- 100289097
- Gene Name
- FSHD region gene 1 family member C, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 20: 28580633 - 28602665 on Build GRCh38
- UniGene ID
- Hs.529357
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132315.1 |
|
6, 7 |
521 |
|
GenBank mRNA |
AK307061.1 |
- |
3, 4 |
400 |
|
- Gene Symbol
- LOC102723780
- Entrez Gene ID
- 102723780
- Gene Name
- protein FRG1-like
- Gene Aliases
- Chromosome Location
- Chr. 22: 10940597 - 10961529 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132320.1 |
|
7, 8 |
793 |
|
- Gene Symbol
- LOC102724813
- Entrez Gene ID
- 102724813
- Gene Name
- protein FRG1B
- Gene Aliases
- Chromosome Location
- Chr. 9|NT_187375: 3219 - 27031 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LOC283788
- Entrez Gene ID
- 283788
- Gene Name
- FSHD region gene 1 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Un|NT_167213: 56349 - 99642 on Build GRCh38
- UniGene ID
- Hs.657131
- Species
- Homo sapiens
View Full Product Details