Gene Transcripts

Gene Symbol
SH2D4B
Entrez Gene ID
387694
Gene Name
SH2 domain containing 4B
Gene Aliases
Chromosome Location
Chr. 10: 80537902 - 80646560 on Build GRCh38
UniGene ID
Hs.147643
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207372.2 NP_997255.2 1 499
XM_017016236.1 XP_016871725.1 1 499
GenBank mRNA AK123885.1 - 1 499

View Full Product Details