Gene Transcripts

Gene Symbol
BSX
Entrez Gene ID
390259
Gene Name
brain specific homeobox
Gene Aliases
BSX1
Chromosome Location
Chr. 11: 122977649 - 122981671 on Build GRCh38
UniGene ID
Hs.449687
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098169.1 NP_001091639.1 1 147
GenBank mRNA AB231738.1 - 1 104

View Full Product Details