Gene Transcripts
- Gene Symbol
- SNHG5
- Entrez Gene ID
- 387066
- Gene Name
- small nucleolar RNA host gene 5
- Gene Aliases
- C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
- Chromosome Location
- Chr. 6: 85677007 - 85678733 on Build GRCh38
- UniGene ID
- Hs.292457
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK311192.1 |
- |
1 |
546 |
|
BC009220.2 |
- |
1 |
515 |
|
BC108291.1 |
- |
1 |
533 |
|
View Full Product Details