Gene Transcripts

Gene Symbol
GLT8D2
Entrez Gene ID
83468
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
Chromosome Location
Chr. 12: 103988983 - 104064177 on Build GRCh38
UniGene ID
Hs.631650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316967.1 NP_001303896.1 2 342
NM_031302.4 NP_112592.1 2 342
XM_011538793.2 XP_011537095.1 1 507
XM_017019999.1 XP_016875488.1 2 588
GenBank mRNA AK056056.1 - 2 348

View Full Product Details