Gene Transcripts
- Gene Symbol
- C5orf47
- Entrez Gene ID
- 133491
- Gene Name
- chromosome 5 open reading frame 47
- Gene Aliases
- Chromosome Location
- Chr. 5: 173989159 - 174007524 on Build GRCh38
- UniGene ID
- Hs.131469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001144954.1 |
NP_001138426.1
|
1, 2 |
421 |
|
XM_011534431.1 |
XP_011532733.1
|
1, 2 |
415 |
|
XM_017009028.1 |
XP_016864517.1
|
1, 2 |
415 |
|
GenBank mRNA |
BC034971.1 |
- |
1, 2 |
387 |
|
BC047225.1 |
- |
1, 2 |
376 |
|
BC053953.1 |
- |
1, 2 |
376 |
|
BC057842.1 |
- |
1, 2 |
362 |
|
BC066964.1 |
- |
1, 2 |
383 |
|
BC092489.1 |
- |
1, 2 |
410 |
|
DB443869.1 |
- |
1, 2 |
421 |
|
View Full Product Details