Gene Transcripts

Gene Symbol
C5orf58
Entrez Gene ID
133874
Gene Name
chromosome 5 open reading frame 58
Gene Aliases
Chromosome Location
Chr. 5: 170231730 - 170252569 on Build GRCh38
UniGene ID
Hs.519749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102609.2 NP_001096079.1 2, 3 185
NM_001305393.1 NP_001292322.1 2, 3 204
NM_001305394.1 NP_001292323.1 2, 3 193
NM_001305395.1 NP_001292324.1 2, 3 185
NM_001305396.1 NP_001292325.1 2, 3 174
NM_001305397.1 NP_001292326.1 2, 3 185
NR_131091.1 2, 3 185
NR_131092.1 1, 2 101
XM_011534432.2 XP_011532734.1 2, 3 656
XM_017009029.1 XP_016864518.1 2, 3 1372
XM_017009030.1 XP_016864519.1 2, 3 1371
XM_017009031.1 XP_016864520.1 2, 3 245
XM_017009032.1 XP_016864521.1 2, 3 169
XM_017009033.1 XP_016864522.1 2, 3 236
GenBank mRNA BC030767.1 - 2, 3 165
BC092511.1 - 1, 2 1165
BU561519.1 - 1, 2 81
BX100176.1 - 1, 2 105
HY005987.1 - 2, 3 204
HY016595.1 - 2, 3 110

View Full Product Details