Gene Transcripts
- Gene Symbol
- SULT1C2P1
- Entrez Gene ID
- 151234
- Gene Name
- sulfotransferase family 1C member 2 pseudogene 1
- Gene Aliases
- SULT1C1P
- Chromosome Location
- Chr. 2: 108322238 - 108353799 on Build GRCh38
- UniGene ID
- Hs.447681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037191.1 |
|
3, 4 |
450 |
|
GenBank mRNA |
AK056906.1 |
- |
3, 4 |
450 |
|
View Full Product Details