Gene Transcripts
- Gene Symbol
- PRELID2
- Entrez Gene ID
- 153768
- Gene Name
- PRELI domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 5: 145587325 - 145835369 on Build GRCh38
- UniGene ID
- Hs.314261
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138492.5 |
NP_612501.3
|
5, 6 |
692 |
|
NM_182960.3 |
NP_892005.1
|
5, 6 |
525 |
|
NM_205846.2 |
NP_995318.1
|
4, 5 |
489 |
|
XM_017009127.1 |
XP_016864616.1
|
5, 6 |
525 |
|
XM_017009128.1 |
XP_016864617.1
|
5, 6 |
525 |
|
XM_017009129.1 |
XP_016864618.1
|
5, 6 |
525 |
|
XM_017009130.1 |
XP_016864619.1
|
5, 6 |
525 |
|
XM_017009131.1 |
XP_016864620.1
|
5, 6 |
525 |
|
GenBank mRNA |
AK095695.1 |
- |
5, 6 |
460 |
|
AK125215.1 |
- |
4, 5 |
424 |
|
BC012033.1 |
- |
5, 6 |
659 |
4476359 |
BC028175.1 |
- |
|
406 |
|
BQ645893.1 |
- |
4, 5 |
459 |
|
BU167299.1 |
- |
4, 5 |
351 |
|
BU186346.1 |
- |
5, 6 |
462 |
|
View Full Product Details