Gene Transcripts
- Gene Symbol
- MDGA2
- Entrez Gene ID
- 161357
- Gene Name
- MAM domain containing glycosylphosphatidylinositol anchor 2
- Gene Aliases
- MAMDC1, c14_5286
- Chromosome Location
- Chr. 14: 46836518 - 47675319 on Build GRCh38
- UniGene ID
- Hs.436380
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113498.2 |
NP_001106970.3
|
12, 13 |
2604 |
|
NM_182830.4 |
NP_878250.2
|
12, 13 |
2310 |
|
XM_011536519.2 |
XP_011534821.1
|
12, 13 |
2459 |
|
XM_011536525.2 |
XP_011534827.1
|
9, 10 |
1741 |
|
XM_017021060.1 |
XP_016876549.1
|
8, 9 |
1570 |
|
GenBank mRNA |
AK309211.1 |
- |
12, 13 |
2607 |
|
AY328482.1 |
- |
6, 7 |
1237 |
|
AY358125.1 |
- |
11, 12 |
2184 |
|
AY369208.1 |
- |
12, 13 |
2400 |
|
BC142947.1 |
- |
12, 13 |
2327 |
|
View Full Product Details